2,8-Bis(trifluoromethyl)quinolin-4-ol 98%

Supplier: Ambeed
Warning

A359094-100G A359094-10G A359094-500G A359094-1G A359094-25G A359094-5G
103826-218EA 11 USD
103826-218 103837-226 76803-442 76803-444 76803-446 76840-390 76840-392 77306-700
2,8-Bis(trifluoromethyl)quinolin-4-ol 98%
2,8-Bis(trifluoromethyl)quinolin-4-ol

Precaución: for Research Use Only

Formula: C₁₁H₅F₆NO
Boiling Pt: 238 °C (760 mm Hg)
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00075091
CAS Number: 35853-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR