2,7-Di-tert-butylpyrene 97%

Supplier: Ambeed
Warning

A510944-5G A510944-100G
103828-396EA 13 USD
103828-396 103839-498 76830-286 77307-658 77886-309
2,7-Di-tert-butylpyrene 97%
2,7-Di-tert-butylpyrene

Precaución: for Research Use Only

Formula: C₂₄H₂₆
Boiling Pt: 448 °C (760 mm Hg)
Flash Pt: 222.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01013641
CAS Number: 24300-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR