2,7-Di-tert-butylfluorene 98%

Supplier: Ambeed
Warning

A539934-25G A539934-5G A539934-10G A539934-500G
103839-746EA 5 USD
103839-746 76843-798 76843-800 77253-676 77253-678
2,7-Di-tert-butylfluorene 98%
2,7-Di-tert-butylfluorene

Precaución: for Research Use Only

Formula: C₂₁H₂₆
MW: 278.43 g/mol
Boiling Pt: 372.4 °C (760 mm Hg)
Melting Pt: 121…124 °C
Flash Pt: 194.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03093998
CAS Number: 58775-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR