2,7-Dimethoxynaphthalene 97%

Supplier: Ambeed
Warning

A245237-1G A245237-5G A245237-10G A245237-25G
103811-628EA 5 USD
103811-628 103851-466 77090-708 77090-710 77249-868 77304-348
2,7-Dimethoxynaphthalene 97%
2,7-Dimethoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₂
MW: 188.23 g/mol
Boiling Pt: 311.2 °C (760 mm Hg)
Melting Pt: 137…139 °C
Flash Pt: 130.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00004064
CAS Number: 3469-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR