2,7-Dihydroxynaphthalene 97%

Supplier: Ambeed
Warning

Synonyms: 2,7-Naphthalenediol, Naphthalene-2,7-diol

A121284-100G A121284-10G A121284-25G A121284-500G A121284-1000G
103808-232EA 233 USD
103808-232 77246-118 77246-120 77246-122 77246-124 77297-822
2,7-Dihydroxynaphthalene 97%
2,7-Dihydroxynaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Melting Pt: 185…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00004085
CAS Number: 582-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR