2,7-Dihydroxy-9-fluorenone 98%

Supplier: Ambeed
Warning

Synonyms: 2,7-Dihydroxy-9H-fluoren-9-one

A151335-100G A151335-10G A151335-500G A151335-1G A151335-25G A151335-5G
103804-248EA 6 USD
103804-248 103842-398 77272-986 77272-988 77272-990 77299-870 77317-944 77317-946
2,7-Dihydroxy-9-fluorenone 98%
2,7-Dihydroxy-9-fluorenone

Precaución: for Research Use Only

Formula: C₁₃H₈O₃
MW: 212.204 g/mol
Boiling Pt: 444.5 °C (760 mm Hg)
Flash Pt: 236.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00037141
CAS Number: 42523-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR