2,7-Dibromopyrene 95%

Supplier: Ambeed
Warning

A210602-1G A210602-5G A210602-100MG A210602-10G A210602-250MG
103818-026EA 107 USD
103818-026 103848-720 77087-098 77087-100 77249-072 77249-074 77249-076
2,7-Dibromopyrene 95%
2,7-Dibromopyrene

Precaución: for Research Use Only

Formula: C₁₆H₈Br₂
Boiling Pt: 469.56 °C (760 mm Hg)
Flash Pt: 277.277 °C
Storage Temperature: Ambient
MDL Number: MFCD09909862
CAS Number: 102587-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR