2,7-Dibromophenanthrene-9,10-dione 97%

Supplier: Ambeed
Warning

A212718-1G A212718-25G A212718-5G A212718-10G
103818-230EA 6 USD
103818-230 103848-926 77087-396 77087-398 77087-400 77249-144
2,7-Dibromophenanthrene-9,10-dione 97%
2,7-Dibromophenanthrene-9,10-dione

Precaución: for Research Use Only

Formula: C₁₄H₆Br₂O₂
MW: 366.01 g/mol
Boiling Pt: 503.8 °C (760 mm Hg)
Flash Pt: 179.1 °C
Storage Temperature: Ambient
MDL Number: MFCD03931078
CAS Number: 84405-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR