2,7-Dibromonaphthalene 98%

Supplier: Ambeed
Warning

A137246-100G A137246-10G A137246-250MG A137246-1G A137246-25G A137246-5G
103802-440EA 15 USD
103802-440 77109-600 77109-602 77109-604 77315-388 77315-390 77315-392
2,7-Dibromonaphthalene 98%
2,7-Dibromonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂
Melting Pt: 139…142 °C
Flash Pt: 184.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00238580
CAS Number: 58556-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR