2,7-Dibromo-9-phenylcarbazole 95%

Supplier: Ambeed
Warning

A100433-10G A100433-250MG A100433-25G A100433-1G A100433-5G
103795-436EA 19 USD
103795-436 103821-132 77093-476 77093-478 77093-480 77282-490 77296-682
2,7-Dibromo-9-phenylcarbazole 95%
2,7-Dibromo-9-phenylcarbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₁Br₂N
Boiling Pt: 505.3 °C (760 mm Hg)
Flash Pt: 259.4 °C
Storage Temperature: Ambient
MDL Number: MFCD28147705
CAS Number: 444796-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR