2,7-Dibromofluorene 97%

Supplier: Ambeed
Warning

Synonyms: 2,7-Dibromo-9H-fluorene

A148664-100G A148664-10G A148664-500G A148664-5G A148664-25G
103842-072EA 5 USD
103842-072 76719-654 76719-656 76719-658 77299-678 77317-534
2,7-Dibromofluorene 97%
2,7-Dibromofluorene

Precaución: for Research Use Only

Formula: C₁₃H₈Br₂
MW: 324.01 g/mol
Boiling Pt: 402.7 °C (760 mm Hg)
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00019048
CAS Number: 16433-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR