2,7-Dibromo-9-fluorenone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,7-Dibromo-9H-fluoren-9-one
A210745-100G
A210745-10G
A210745-1G
A210745-25G
A210745-5G
103818-036EA
5
USD
103818-036
103848-730
76999-606
76999-608
77303-142
77303-144
77303-146
2,7-Dibromo-9-fluorenone 98%
2,7-Dibromo-9-fluorenone
Precaución: for Research Use Only
Formula:
C₁₃H₆Br₂O MW: 338 g/mol Melting Pt: 203…205 °C Flash Pt: 163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010790 CAS Number: 14348-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...