2,7-Dibromocarbazole 98%

Supplier: Ambeed
Warning

A176958-1G A176958-25G A176958-5G A176958-100G A176958-10G A176958-250MG A176958-500G
103807-190EA 7 USD
103807-190 103845-516 77082-880 77082-882 77082-884 77248-164 77248-166 77248-168 77746-153
2,7-Dibromocarbazole 98%
2,7-Dibromocarbazole

Precaución: for Research Use Only

Formula: C₁₂H₇Br₂N
Boiling Pt: 459 °C (760 mm Hg)
Melting Pt: 225…230 °C
Flash Pt: 231.4 °C
Storage Temperature: Ambient
MDL Number: MFCD09033507
CAS Number: 136630-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR