2,7-Dibromo-9,9'-spirobifluorene 98%
Supplier: Ambeed
|
Warning
|
A264175-100G
A264175-10G
A264175-1G
A264175-25G
A264175-5G
103812-656EA
6
USD
103812-656
103852-548
77001-000
77001-002
77304-884
77304-886
77304-888
2,7-Dibromo-9,9'-spirobifluorene 98%
2,7-Dibromo-9,9'-spirobifluorene
Precaución: for Research Use Only
Formula:
C₂₅H₁₄Br₂ Boiling Pt: 557.4 °C (760 mm Hg) Melting Pt: 334…336 °C Flash Pt: 335.129 °C Storage Temperature: Ambient |
MDL Number:
MFCD08704218 CAS Number: 171408-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...