2,7-Dibromo-9,9-dihexylfluorene 97%

Supplier: Ambeed
Warning

Synonyms: 2,7-Dibromo-9,9-dihexyl-9H-fluorene

A362895-25G A362895-5G A362895-100G A362895-10G A362895-1G
103826-332EA 7 USD
103826-332 103837-348 76840-566 76840-568 77003-394 77003-396 77306-746
2,7-Dibromo-9,9-dihexylfluorene 97%
2,7-Dibromo-9,9-dihexylfluorene

Precaución: for Research Use Only

Formula: C₂₅H₃₂Br₂
MW: 492.34 g/mol
Boiling Pt: 522.2 °C (760 mm Hg)
Melting Pt: 67…71 °C
Flash Pt: 312.3 °C
Storage Temperature: Ambient
MDL Number: MFCD03427215
CAS Number: 189367-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR