2,6-Di-tert-butylpyridine 98%

Supplier: Ambeed
Warning

A287478-1G A287478-25G A287478-5G A287478-100G A287478-10G
103813-758EA 11 USD
103813-758 103834-900 76837-452 76837-454 76837-456 77114-200 77114-202
2,6-Di-tert-butylpyridine 98%
2,6-Di-tert-butylpyridine

Precaución: for Research Use Only

Formula: C₁₃H₂₁N
Boiling Pt: 208.5 °C (760 mm Hg)
Melting Pt: 2 °C
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00006306
CAS Number: 585-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR