2,6-Di-tert-butyl-4-vinylphenol 95%

Supplier: Ambeed
Warning

A197129-100MG A197129-250MG
103816-894EA 330 USD
103816-894 103847-512 77248-756 77248-758
2,6-Di-tert-butyl-4-vinylphenol 95%
2,6-Di-tert-butyl-4-vinylphenol

Precaución: for Research Use Only

Formula: C₁₆H₂₄O
MW: 232.36 g/mol
Melting Pt: 51…51.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD28505064
CAS Number: 19263-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR