2,4-Dimethyl-6-hydroxypyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethylpyrimidin-4-ol, 2,6-Dimethyl-4-pyrimidinol, 6-Hydroxy-2,4-dimethylpyrimidine

A143585-100G A143585-10G A143585-500G A143585-25G A143585-5G
77109-900EA 141.1 USD
77109-900 77109-902 77109-904 77316-580 77316-582
2,4-Dimethyl-6-hydroxypyrimidine 98%
2,4-Dimethyl-6-hydroxypyrimidine

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Melting Pt: 198…200 °C
Flash Pt: 75.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00006105
CAS Number: 6622-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR