4-Amino-2,6-dimethylpyrimidine 98%

Supplier: Ambeed
Warning

A296440-1G A296440-25G A296440-5G A296440-100G A296440-10G A296440-250MG
103814-100EA 9 USD
103814-100 103835-242 76837-910 76837-912 76837-914 76977-654 76977-656 76977-658
4-Amino-2,6-dimethylpyrimidine 98%
4-Amino-2,6-dimethylpyrimidine

Precaución: For research use only

Formula: C₆H₉N₃
MW: 123.16 g/mol
Boiling Pt: 238.7 °C (760 mm Hg)
Melting Pt: 182…185 °C
Flash Pt: 121.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00006106
CAS Number: 461-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR