2,6-Dimethylpyridin-4-ol 95%
Supplier: Ambeed
|
Warning
|
A150121-100G
A150121-10G
103842-264EA
12
USD
103842-264
77272-962
77272-964
2,6-Dimethylpyridin-4-ol 95%
2,6-Dimethylpyridin-4-ol
Precaución: for Research Use Only
Formula:
C₇H₉NO Boiling Pt: 351 °C (760 mm Hg) Melting Pt: 230…232 °C Flash Pt: 93.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00956021 CAS Number: 13603-44-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...