4-Amino-2,6-dimethylpyridine 97%
Supplier: Ambeed
|
Warning
|
A244859-10G
A244859-250MG
A244859-1G
A244859-5G
103811-604EA
16
USD
103811-604
103851-438
76987-728
76987-730
77090-656
77090-658
4-Amino-2,6-dimethylpyridine 97%
4-Amino-2,6-dimethylpyridine
Precaución: for Research Use Only
Formula:
C₇H₁₀N₂ Flash Pt: 126.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00130078 CAS Number: 3512-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...