3-Amino-2,6-dimethylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethylpyridin-3-amine

A160547-100G A160547-10G A160547-1G A160547-25G A160547-5G
103805-394EA 5 USD
103805-394 103843-626 77110-648 77110-650 77319-570 77319-572 77319-574
3-Amino-2,6-dimethylpyridine 98%
3-Amino-2,6-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 231.5 °C (760 mm Hg)
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD01646409
CAS Number: 3430-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR