2,6-Dimethylpiperazine 98%

Supplier: Ambeed
Danger

A575182-25G A575182-5G A575182-10G A575182-1G
103839-976EA 8 USD
103839-976 76844-106 76844-108 77157-370 77157-372
2,6-Dimethylpiperazine 98%
2,6-Dimethylpiperazine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 114.19
Boiling Pt: 162 °C (760 mm Hg)
Melting Pt: 108…111 °C
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00005956
CAS Number: 108-49-6
UN: 1325
ADR: 4.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR