2,6-Dimethylisonicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethylpyridine-4-carboxylic acid

A145681-100MG A145681-250MG A145681-1G A145681-5G
103803-572EA 83 USD
103803-572 103841-654 77247-110 77247-112 77299-470 77566-964
2,6-Dimethylisonicotinic acid 98%
2,6-Dimethylisonicotinic acid

Precaución: For research use only.

Formula: C₈H₉NO₂
MW: 151.163 g/mol
Boiling Pt: 353.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03731176
CAS Number: 54221-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR