2,6-Dimethylbenzoic acid 98%

Supplier: Ambeed
Warning

A152671-100G A152671-10G A152671-500G A152671-1G A152671-25G A152671-5G
103804-414EA 6 USD
103804-414 103842-586 76689-406 76689-408 76689-410 77318-208 77318-210 77318-212
2,6-Dimethylbenzoic acid 98%
2,6-Dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 272.7 °C (760 mm Hg)
Melting Pt: 114…116 °C
Flash Pt: 126.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00002483
CAS Number: 632-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR