2,6-Dimethylbenzaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 2-Formyl-m-xylene

A150873-100G A150873-10G A150873-1G A150873-25G A150873-5G
103804-208EA 5 USD
103804-208 103842-348 77247-240 77247-242 77299-830 77317-882 77317-884
2,6-Dimethylbenzaldehyde 95%
2,6-Dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 230 °C (760 mm Hg)
Melting Pt: 27…30 °C
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00128003
CAS Number: 1123-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR