2,6-Dimethyl-3-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethylpyridin-3-ol, 3-Hydroxy-2,6-lutidine

A100550-10G A100550-250MG A100550-25G A100550-1G A100550-5G
103795-506EA 18 USD
103795-506 103821-196 76765-110 76765-112 76765-114 77296-750 77296-752
2,6-Dimethyl-3-hydroxypyridine 98%
2,6-Dimethyl-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₇H₉NO
MW: 123.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00228692
CAS Number: 1122-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR