2,4-Dimethoxypyrimidine-6-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethoxypyrimidine-4-carboxylic acid

A156694-250MG A156694-1G A156694-25G A156694-5G
103804-912EA 55 USD
103804-912 103843-114 76768-386 77318-910 77318-912 77318-914
2,4-Dimethoxypyrimidine-6-carboxylic acid 97%
2,4-Dimethoxypyrimidine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₄
MW: 184.15 g/mol
Boiling Pt: 396.1 °C (760 mm Hg)
Melting Pt: 162 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01568105
CAS Number: 59864-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR