2,6-Dimethoxypyridine 98%

Supplier: Ambeed
Warning

A187712-25G A187712-10G A187712-1G A187712-5G A187712-500G
103846-632EA 6 USD
103846-632 77084-346 77138-960 77138-962 77302-094 77567-750
2,6-Dimethoxypyridine 98%
2,6-Dimethoxypyridine

Precaución: For research use only.

Formula: C₇H₉NO₂
MW: 139.15 g/mol
Boiling Pt: 178…180 °C (760 mm Hg)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00006266
CAS Number: 6231-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR