2,6-Dimethoxy-3-pyridineboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethoxypyridine-3-boronic acid, 2,6-Dimethoxypyridin-3-ylboronic acid

A205168-1G A205168-25G A205168-10G A205168-250MG A205168-5G A205168-100G
103817-610EA 7 USD
103817-610 103848-276 77086-474 77086-476 77112-298 77112-300 77302-948 77685-524
2,6-Dimethoxy-3-pyridineboronic acid 97%
2,6-Dimethoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₄
Boiling Pt: 341.2 °C (760 mm Hg)
Melting Pt: 128…130 °C
Flash Pt: 160.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03788239
CAS Number: 221006-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR