2,6-Dimethoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A471853-5G A471853-100G A471853-10G A471853-1G A471853-500G A471853-25G
103839-108EA 8 USD
103839-108 76843-014 77196-692 77196-694 77196-696 77196-698 77307-490
2,6-Dimethoxyphenylboronic acid 98%
2,6-Dimethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
Boiling Pt: 372.8 °C (760 mm Hg)
Melting Pt: 110…112 °C
Flash Pt: 179.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01318987
CAS Number: 23112-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR