2,6-Dimethoxybenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A210598-100G
A210598-10G
A210598-1G
A210598-25G
A210598-5G
A210598-500G
103818-024EA
5
USD
103818-024
103848-718
77097-660
77097-662
77303-134
77303-136
77303-138
77685-550
2,6-Dimethoxybenzaldehyde 98%
2,6-Dimethoxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₉H₁₀O₃ Boiling Pt: 210 °C (760 mm Hg) Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010862 CAS Number: 3392-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...