2,6-Dimethoxyaniline 97%

Supplier: Ambeed
Warning

A294710-25G A294710-100G A294710-10G A294710-250MG A294710-1G A294710-5G
103814-020EA 6 USD
103814-020 103835-160 76837-800 77166-870 77166-872 77166-874 77305-660 77305-662
2,6-Dimethoxyaniline 97%
2,6-Dimethoxyaniline

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
Boiling Pt: 253.6 °C (760 mm Hg)
Melting Pt: 71…74 °C
Flash Pt: 115.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00053934
CAS Number: 2734-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR