4-Hydroxy-2,6-dimethoxybenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethoxy-4-hydroxybenzaldehyde

A204806-1G A204806-25G A204806-5G A204806-10G A204806-250MG
103817-566EA 27 USD
103817-566 103848-232 77086-414 77086-416 77086-418 77248-906 77248-908
4-Hydroxy-2,6-dimethoxybenzaldehyde 98%
4-Hydroxy-2,6-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.1733 g/mol
Boiling Pt: 370.4 °C (760 mm Hg)
Melting Pt: 225 °C
Flash Pt: 152.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01407645
CAS Number: 22080-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR