2,6-Diisopropylaniline 97%
Supplier: Ambeed
|
Warning
|
A286085-25G
A286085-100G
A286085-10G
A286085-500G
103809-948EA
71
USD
103809-948
76837-366
77099-248
77099-250
77099-252
2,6-Diisopropylaniline 97%
2,6-Diisopropylaniline
Precaución: for Research Use Only
Formula:
C₁₂H₁₉N Boiling Pt: 420.7 °C (760 mm Hg) Melting Pt: 45 °C Flash Pt: 117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008887 CAS Number: 24544-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...