2,6-Dihydroxy-9,10-anthraquinone 97%

Supplier: Ambeed
Warning

A173241-1G A173241-5G A173241-100G A173241-10G A173241-250MG
103806-806EA 13 USD
103806-806 103845-100 77082-346 77082-348 77248-098 77248-100 77248-102
2,6-Dihydroxy-9,10-anthraquinone 97%
2,6-Dihydroxy-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₈O₄
Boiling Pt: 442.1 °C (760 mm Hg)
Melting Pt: 235.3 °C
Flash Pt: 235.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00001228
CAS Number: 84-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR