2,6-Difluoro-3-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluoropyridine-3-boronic acid

A169507-10G A169507-250MG A169507-1G A169507-25G A169507-5G
103806-390EA 17 USD
103806-390 103844-650 76985-342 76985-344 77081-734 77081-736 77081-738
2,6-Difluoro-3-pyridineboronic acid 98%
2,6-Difluoro-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BF₂NO₂
MW: 158.90 g/mol
Boiling Pt: 314.8 °C (760 mm Hg)
Flash Pt: 144.161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788245
CAS Number: 136466-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR