4-Amino-2,6-difluoropyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluoropyridin-4-amine

A212346-100MG A212346-250MG
103818-192EA 56 USD
103818-192 103848-890 76825-934 77550-366
4-Amino-2,6-difluoropyridine 97%
4-Amino-2,6-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₄F₂N₂
MW: 130.098 g/mol
Melting Pt: 124…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08436089
CAS Number: 63489-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR