2,6-Difluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Dihydroxy(2,6-difluorophenyl)borane, 2,6-Difluorobenzeneboronic acid, (2,6-Difluorophenyl)metaboric acid

A308353-100G A308353-10G A308353-1G A308353-25G A308353-5G
103814-552EA 5 USD
103814-552 103835-728 77290-128 77290-130 77305-982 77305-984 77305-986
2,6-Difluorophenylboronic acid 98%
2,6-Difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
Boiling Pt: 260.1 °C (760 mm Hg)
Melting Pt: 147…149 °C
Flash Pt: 87.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00792436
CAS Number: 162101-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR