2,6-Difluoropyridine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluoroisonicotinic acid

A315491-1G A315491-5G A315491-100MG A315491-10G A315491-250MG A315491-25G
103825-074EA 71 USD
103825-074 103835-998 76838-826 76838-828 77251-048 77251-050 77251-052 77251-054
2,6-Difluoropyridine-4-carboxylic acid 97%
2,6-Difluoropyridine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 398.6 °C (760 mm Hg)
Flash Pt: 194.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09864665
CAS Number: 88912-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR