2,6-Difluorobenzonitrile 98%

Supplier: Ambeed
Danger

A136814-100G A136814-10G A136814-500G A136814-1000G A136814-25G
103808-542EA 86 USD
103808-542 76996-684 76996-686 76996-688 77315-272 77315-274
2,6-Difluorobenzonitrile 98%
2,6-Difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 174 °C (760 mm Hg)
Melting Pt: 25…28 °C
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00001778
CAS Number: 1897-52-5
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR