2,6-Difluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A189464-100G A189464-10G A189464-500G A189464-5G A189464-1G A189464-25G
103816-188EA 8 USD
103816-188 103846-780 76769-836 76769-838 76769-840 77084-554 77302-176 77302-178
2,6-Difluorobenzaldehyde 98%
2,6-Difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 193.6 °C (760 mm Hg)
Melting Pt: 15…17 °C
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010293
CAS Number: 437-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR