2,6-Difluoro-4-methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

A169670-250MG A169670-1G A169670-25G A169670-5G
103806-398EA 15 USD
103806-398 103844-662 77028-978 77081-752 77081-754 77081-756
2,6-Difluoro-4-methoxyphenylboronic acid 97%
2,6-Difluoro-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
MW: 187.94 g/mol
Melting Pt: 125…130 °C
Flash Pt: 116.7 °C
Storage Temperature: Ambient
MDL Number: MFCD05664299
CAS Number: 406482-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR