2,6-Difluoro-4-methoxyaniline 98%

Supplier: Ambeed
Warning

A356404-10G A356404-250MG A356404-1G A356404-25G A356404-5G
103826-170EA 34 USD
103826-170 103837-178 76788-464 76788-466 76840-310 76840-312 76840-314
2,6-Difluoro-4-methoxyaniline 98%
2,6-Difluoro-4-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇F₂NO
Boiling Pt: 172.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04115922
CAS Number: 151414-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR