2,6-Difluoro-4-iodopyridine 97%

Supplier: Ambeed
Warning

A457089-10G A457089-250MG A457089-1G A457089-25G A457089-5G A457089-100G
103827-870EA 17 USD
103827-870 103838-946 76818-392 76818-394 76842-810 76842-812 76842-814 77613-414
2,6-Difluoro-4-iodopyridine 97%
2,6-Difluoro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂F₂IN
MW: 240.977 g/mol
Boiling Pt: 222 °C (760 mm Hg)
Melting Pt: 38…40 °C
Flash Pt: 88.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11977432
CAS Number: 685517-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR