2,6-Difluoro-4-hydroxybenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: 3,5-Difluoro-4-(hydroxymethyl)phenol

A205678-10G A205678-1G A205678-25G A205678-5G
103817-656EA 33 USD
103817-656 103848-320 76975-334 77086-544 77086-546 77086-548
2,6-Difluoro-4-hydroxybenzyl alcohol 95%
2,6-Difluoro-4-hydroxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆F₂O₂
MW: 160.1200 g/mol
Boiling Pt: 286 °C (760 mm Hg)
Melting Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD11040173
CAS Number: 438049-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR