2,6-Difluoro-4-hydroxybenzonitrile 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Cyano-3,5-difluorophenol
A358241-10G
A358241-1G
A358241-5G
103826-206EA
6
USD
103826-206
103837-212
76773-334
76840-376
76840-378
2,6-Difluoro-4-hydroxybenzonitrile 98%
2,6-Difluoro-4-hydroxybenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₃F₂NO MW: 155.1 g/mol Boiling Pt: 210.2 °C (760 mm Hg) Melting Pt: 127 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094499 CAS Number: 123843-57-2 UN: 3439 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...