2,6-Difluoro-4-hydroxybenzaldehyde 98%

Supplier: Ambeed
Warning

A194628-10G A194628-250MG A194628-1G A194628-5G A194628-25G
103816-674EA 25 USD
103816-674 103847-284 77029-930 77029-932 77085-210 77085-212 77302-424
2,6-Difluoro-4-hydroxybenzaldehyde 98%
2,6-Difluoro-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O₂
Boiling Pt: 252.6 °C (760 mm Hg)
Melting Pt: 179…182 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07772042
CAS Number: 532967-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR