2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzoic acid

A124916-100MG A124916-250MG A124916-1G A124916-5G
103800-594EA 89 USD
103800-594 76810-778 76810-780 77298-026 77298-028
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid 98%
2,6-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅BF₂O₄
MW: 284.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16996277
CAS Number: 1008119-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR