2,6-Difluoro-3-nitroaniline 97%

Supplier: Ambeed
Warning

A219193-10G A219193-250MG A219193-1G A219193-5G
103818-768EA 9 USD
103818-768 103849-642 76814-830 76814-832 77088-186 77088-188
2,6-Difluoro-3-nitroaniline 97%
2,6-Difluoro-3-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄F₂N₂O₂
Boiling Pt: 285.5 °C (760 mm Hg)
Flash Pt: 126.4 °C
Storage Temperature: Ambient
MDL Number: MFCD11110684
CAS Number: 25892-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR